Learn More
Enzo Life Sciences UbcH12 (human), (recombinant) (His-tag) (100 µg)
SDP
Catalog No. 502012303
Encompass_Preferred
Purity: ≥95% (SDS-PAGE). Formulation: Liquid. In HEPES, pH 7.5, containing 100mM NaCl, 1mM DTT, 10% glycerol. Source: Produced in E. coli BL21 (λDE3). UbcH12 is fused to an N-terminal His-tag. Long Term Storage: -80°C.
Catalog No. 50-201-2303 Supplier Enzo Life Sciences Supplier No. BMLUW91450100
Purity: ≥95% (SDS-PAGE). Formulation: Liquid. In HEPES, pH 7.5, containing 100mM NaCl, 1mM DTT, 10% glycerol. Source: Produced in E. coli BL21 (λDE3). UbcH12 is fused to an N-terminal His-tag. Long Term Storage: -80°C.