Learn More
Enzo Life Sciences UbcH13/Mms2 (human), (recombinant) (His-tag) (100 µg)
SDP
Supplier:  Enzo Life Sciences BMLUW95650100
Encompass_Preferred
A heterodimer composed of the catalytically active ubiquitin-conjugating enzyme UbcH13 and its catalytically inactive paralog hMms2, which lacks an active site cysteine residue. A heterodimer composed of the catalytically active ubiquitin-conjugating enzyme UbcH13 and its catalytically inactive paralog hMms2, which lacks an active site cysteine residue. Purity: ≥95% (SDS-PAGE). Formulation: Liquid. In 50mM HEPES, pH 8.0, 50mM sodium chloride, 10% glycerol, 1mM DTT. Source: Produced in E. coli. Long Term Storage: -80°C.
Catalog No. 50-201-2317