Learn More
Enzo Life Sciences UbcH5b (human), (recombinant) (His-tag) (1 mg)
SDP
Supplier:  Enzo Life Sciences BMLUW90601000
Encompass_Preferred
Purity: ≥90% (SDS-PAGE). Formulation: Liquid. In 25mM TRIS-HCl, pH 7.5, containing 150mM NaCl and 0.5mM DTT. Source: Produced in E. coli. Long Term Storage: -80°C.
Catalog No. 50-201-2298