Learn More
Enzo Life Sciences UbcH5b (human), (recombinant) (His-tag) (100 µg)
SDP
Supplier:  Enzo Life Sciences BMLUW90600100
Encompass_Preferred
MW: ~18kDa. Purity: ≥90% (SDS-PAGE). Formulation: Liquid. In 25mM TRIS-HCl, pH 7.5, containing 150mM NaCl and 0.5mM DTT. Source: Produced in E. coli. Use/Stability: Enzyme is stable to multiple freeze/thaw cycles. UniProt: P62837. Long Term Storage: -80°C
Catalog No. NC1427343