Learn More
Enzo Life Sciences UbcH5c (human), (recombinant) (His-tag) (100 µg)
SDP
Supplier:  Enzo Life Sciences BMLUW90700100
Encompass_Preferred
Purity: ≥90% (SDS-PAGE). Formulation: Liquid. In TRIS-HCl, pH 7.5, containing 1mM DTT. Source: Produced in E. coli. Long Term Storage: -80°C.
Catalog No. 50-201-2300