Learn More
Enzo Life Sciences UbcH6 (human), (recombinant) (His-tag) (100 µg)
SDP
Supplier:  Enzo Life Sciences BMLUW87100100
Encompass_Preferred
Purity: ≥95% (SDS-PAGE). Formulation: Liquid. In PBS, pH 7.2, containing 1mM DTT and 10% glycerol. Source: Produced in E. coli. Long Term Storage: -80°C.
Catalog No. 50-201-2291