Learn More
Sino Biological UBCH8 / UBE2L6 Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 12641RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human UBCH8 / UBE2L6 (rh UBCH8 / UBE2L6; Catalog#12641-H07E; O14933-1; Met1-Ser153). UBCH8 / UBE2L6 specific IgG was purified by Human UBCH8 / UBE2L6 affinity chromatography.
Catalog No. 50-224-7935