Learn More
Medchemexpress LLC HY-115453 10mg Medchemexpress, UBCS039 CAS:358721-70-7 Purity:>98%
SDP
Catalog No. 501969383
Encompass_Preferred
Medchemexpress, HY-115453 10mg UBCS039 CAS:358721-70-7 UBCS039 is the first synthetic, specific Sirtuin 6 (SIRT6) activator, inducing autophagy in human tumor cells, with an EC50 of 38 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9383 Supplier Medchemexpress LLC Supplier No. HY11545310MG
Medchemexpress, HY-115453 10mg UBCS039 CAS:358721-70-7 UBCS039 is the first synthetic, specific Sirtuin 6 (SIRT6) activator, inducing autophagy in human tumor cells, with an EC50 of 38 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.