Learn More
Selleck Chemical LLC UBCS039, Solution 10mM (1mL in DMSO)
SDP
Catalog No. NC2937819
Encompass_Preferred
UBCS039 is the first synthetic SIRT6 activator with EC50 of 38 μM,induces a time-dependent activation of autophagy in several human tumor cell lines.
Catalog No. NC2937819 Supplier Selleck Chemical LLC Supplier No. S890910MM/1ML
UBCS039 is the first synthetic SIRT6 activator with EC50 of 38 μM,induces a time-dependent activation of autophagy in several human tumor cell lines.