Learn More
Aviva Systems Biology Corp UBE2C Antibody - middle region (ARP43474_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43474_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against UBE2C. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human UBE2C. Alternate names: UBCH10, dJ447F3.2ApplicationWB. Species reactivity: Human
Catalog No. 50-285-076