Learn More
Aviva Systems Biology Corp UBE2D3 Antibody - N-terminal region (ARP43009_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43009_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against UBE2D3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human UBE2D3. Alternate names: E2(17)KB3, MGC43926, MGC5416, UBC4/5, UBCH5CApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-58391