Learn More
Aviva Systems Biology Corp UBE2E1 Antibody - N-terminal region (ARP43476_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43476_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against UBE2E1. It was validated on Western Blot. . Alternate names: UBCH6ApplicationWB. Species reactivity: Dog, Human, Mouse, Rabbit
Catalog No. 50-285-077