Learn More
Aviva Systems Biology Corp UBE2F Antibody - N-terminal region (ARP52515_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52515_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against UBE2F. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human UBE2F. Alternate names: MGC18120, NCE2ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53694