Learn More
Aviva Systems Biology Corp Ube2g1 Antibody - N-terminal region (ARP43014_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43014_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Ube2g1. It was validated on Western Blot. . Alternate names: 2700059C12Rik, AI256795, AU014992, AW552068, D130023C12RikApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-285-051