Learn More
Aviva Systems Biology Corp UBE2I Antibody - middle region (ARP43022_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43022_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against UBE2I. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human UBE2I. Alternate names: C358B7.1, P18, UBC9ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-58395