Learn More
Aviva Systems Biology Corp UBE2L3 Antibody - C-terminal region (ARP43026_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43026_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against UBE2L3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human UBE2L3. Alternate names: E2-F1, L-UBC, UBCH7, UbcM4ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58398