Learn More
Aviva Systems Biology Corp UBE2L6 Antibody - middle region (ARP43034_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43034_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against UBE2L6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human UBE2L6. Alternate names: MGC40331, RIG-B, UBCH8ApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-58403