Learn More
Aviva Systems Biology Corp UBE2N Antibody - middle region (ARP34089_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34089_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against UBE2N. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human UBE2N. Alternate names: UBC13, UbcH-benApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Yeast, Zebrafish
Catalog No. 50-014-55243