Learn More
Sino Biological Recombinant Human UBE2E3/UBCH9 Protein (Full Length, His Tag), Active
Catalog No. 502572669
Encompass_Preferred
Recombinant full length human UBE2E3 was expressed in E. coli cells using an N-terminal His tag.
Catalog No. 50-257-2669 Supplier Sino Biological Supplier No. U219380H1000
Add to Cart
Add to Cart
Request a Quote
Recombinant full length human UBE2E3 was expressed in E. coli cells using an N-terminal His tag.