Learn More
Aviva Systems Biology Corp UBE2Q1 Antibody - middle region (ARP62942_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62942_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against UBE2Q1. It was validated on Western Blot. . Alternate names: GTAP, NICE-5, PRO3094, UBE2QApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast
Catalog No. 50-287-816