Learn More
LifeSensors UBE2R1 (CDC34), His6 3nmol
SDP
Catalog No. NC3779494
Encompass_Preferred
236 amino acid product of the CDC34 gene. It controls cellular proliferation through regulation of p27/Kip1 protein levels. Blocking CDC34 activity enhances the anti-myeloma effects of Bortezomib. Useful for UPS research
Catalog No. NC3779494 Supplier LifeSensors Supplier No. UB208H0030
236 amino acid product of the CDC34 gene. It controls cellular proliferation through regulation of p27/Kip1 protein levels. Blocking CDC34 activity enhances the anti-myeloma effects of Bortezomib. Useful for UPS research