Learn More
Aviva Systems Biology Corp UBE2T Antibody - N-terminal region (ARP43145_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43145_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against UBE2T. It was validated on Western Blot. UBE2T,ubiquitin-conjugating enzyme E2T (putative),PIG50; HSPC150; UBE2T,ubiquitin-conjugating enzyme E2 T,ubiquitin-conjugating enzyme E2 T,ubiquitin-protein ligase T,ubiquitin carrier protein T,ubiquitin conjugating enzyme,cell proliferation-inducing gene 50 protein,HSPC150 protein similar to ubiquitin-conjugating enzymeThe immunogen is a synthetic peptide directed towards the N terminal region of human UBE2T. Alternate names: HSPC150, PIG50ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-285-056