Learn More
Aviva Systems Biology Corp UBE2V1 Antibody - N-terminal region (ARP61717_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61717_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against UBE2V1. It was validated on Western Blot. . Alternate names: CIR1, CROC-1, CROC1, UBE2V, UEV-1, UEV1, UEV1AApplicationWB. Species reactivity: Human
Catalog No. 50-287-448