Learn More
Aviva Systems Biology Corp UBE2V2 Antibody - C-terminal region (ARP64750_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64750_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against UBE2V2. It was validated on Western Blot. . Alternate names: DDVIT1, DDVit-1, EDAF-1, EDPF-1, EDPF1, MMS2, UEV-2, UEV2ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-288-356