Learn More
Aviva Systems Biology Corp UBE3A Antibody - N-terminal region (ARP42950_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42950_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against UBE3A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human UBE3A. Alternate names: ANCR, AS, E6-AP, EPVE6AP, FLJ26981, HPVE6AApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-58370