Learn More
Aviva Systems Biology Corp UBE3B Antibody - middle region (ARP43387_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43387_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against UBE3B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human UBE3B. Alternate names: DKFZp586K2123, DKFZp686A1051, FLJ45294, MGC131858, MGC78388ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58516