Learn More
Aviva Systems Biology Corp UBE4B Antibody - N-terminal region (ARP54552_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54552_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against UBE4B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human UBE4B. Alternate names: E4, HDNB1, KIAA0684, UBOX3, UFD2, UFD2AApplicationWB. Species reactivity: Dog, Human, Pig
Catalog No. 50-014-61242