Learn More
Selleck Chemical LLC Bestatin 10mg 58970-76-6 Ubenimex
SDP
Catalog No. 507225
Encompass_Preferred
Bestatin (Ubenimex) is a potent aminopeptidase-B and leukotriene (LT) A4 hydrolase inhibitor, used in the treatment of acute myelocytic leukemia. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-722-5 Supplier Selleck Chemical LLC Supplier No. S159110MG
Bestatin (Ubenimex) is a potent aminopeptidase-B and leukotriene (LT) A4 hydrolase inhibitor, used in the treatment of acute myelocytic leukemia. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.