Learn More
LifeSensors Ubiquilabel™ – [N-15 (98%)] 10mg
SDP
Catalog No. NC3728757
Encompass_Preferred
15N, heavy-labeled ubiquitin provides a stable standard for ubiquitin structural study by NMR. The recombinant human ubiquitin is purified from E. coli expressing the human ubiquitin gene. The cells are grown in media where the sole nitrogen source is 15N labeled ammonium sulfate. The recombinant human ubiquitin is purified without heating by a proprietary procedure that preserves the native structure of the protein. The purified protein is homogenous when analyzed by overloaded SDS gels. The pools of purified protein solution are extensively dialyzed against water and lyophilized.
Catalog No. NC3728757 Supplier LifeSensors Supplier No. NS01010010
15N, heavy-labeled ubiquitin provides a stable standard for ubiquitin structural study by NMR. The recombinant human ubiquitin is purified from E. coli expressing the human ubiquitin gene. The cells are grown in media where the sole nitrogen source is 15N labeled ammonium sulfate. The recombinant human ubiquitin is purified without heating by a proprietary procedure that preserves the native structure of the protein. The purified protein is homogenous when analyzed by overloaded SDS gels. The pools of purified protein solution are extensively dialyzed against water and lyophilized.