Learn More
Cayman Chemical UbIquInon asy Reagent 100uL
SDP
Catalog No. 502892657
Encompass_Preferred
An alkaloid with enzyme inhibitory activity an inhibitor of AChE BChE and prolyl endopeptidase (IC50s 281 911 and 907 UM respectively)
Catalog No. 50-289-2657 Supplier Cayman Chemical Supplier No. 700018100 UL
An alkaloid with enzyme inhibitory activity an inhibitor of AChE BChE and prolyl endopeptidase (IC50s 281 911 and 907 UM respectively)