Learn More
Enzo Life Sciences Ubiquinone-10 (2mg). CAS: 606-06-4
SDP
Supplier:  Enzo Life Sciences ALX270295M002
Encompass_Preferred
Inhibitor of the mitochondrial permeability transition pore (MPTP). This inhibitory effect can be counteracted by ubiquinone-5 (ALX-270-294). Alternative name: Coenzyme Q2, 2,3-Dimethoxy-5-methyl-geranyl-1,4-benzoquinone. Formula: C19H26O4. MW: 318.4. Purity: ≥90% (HPLC). Appearance: Red liquid. Solubility: Soluble in acetone (50mg/ml). Long Term Storage: -20°C.
Catalog No. 50-200-8474