Learn More
Enzo Life Sciences Ubiquinone-5 (2mg). CAS: 727-81-1
SDP
Supplier:  Enzo Life Sciences ALX270294M002
Encompass_Preferred
Counteracts inhibitory effects on mitochondrial permeability transition pore (MPTP) of decylubiquinone (BML-CM115) and ubiquinone-10 (ALX-270-295). Alternative name: Coenzyme Q1, 2,3-Dimethoxy-5-methyl-6-(3-methyl-2-butenyl)-1,4-benzoquinone. Formula: C14H18O4. MW: 250.3. Purity: ≥95% (HPLC). Appearance: Orange-red to red liquid. Solubility: Soluble in acetone (50mg/ml). Long Term Storage: -20°C.
Catalog No. 50-200-8473