Learn More
Ubpbio High purity recombinant human wild type ubiquitin
Supplier:  Ubpbio E1100
Encompass_Preferred
Ubiquitin (Ub) is a 76 amino acid protein widely expressed in the cytoplasmic and nucleus of cells. Ub is posttranslationally conjugated to proteins by the E1, E2, E3 protein ubiquitination cascade. Ub can be conjugated on proteins as monoUb or polyUb chains. Protein ubiquitination plays both proteolytic and nonproteolytic functions. Usually, polyubiquitinated proteins are targeted to the 26S proteasome for proteolysis. Typical concentration to support in vitro ubiquitination is 50-100 µM.
Catalog No. 50-233-9265