Learn More
Novus Biologicals™ Ubiquitin-activating Enzyme/UBE1 Recombinant Protein Antigen

Description
- A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human Ubiquitin-activating Enzyme/UBE1
- This peptide is useful as a blocking peptide for NBP2-76500.
- Protein was purified by IMAC chromatography.
- The expected concentration is greater than 0.5 mg/mL
- This product is intended to be used as a blocking antigen for antibody competition assays. Any other use of this antigen is done at the risk of the user. The use of this product for commercial production is strictly prohibited. Please contact technical support if you have any questions.
- This product is for research use only and is not approved for use in humans or in clinical diagnosis.
Specifications
Specifications
| Gene ID (Entrez) | 7317 |
| Purification Method | >80% by SDS-PAGE and Coomassie blue staining |
| Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
| For Use With (Application) | Antibody Competition |
| Gene Symbol | UBA1 |
| Molecular Weight (g/mol) | 35 kDa |
| Quantity | 100 μL |
| Source | E. coli |
| Specific Reactivity | This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-54955. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml |
| Storage Buffer | PBS and 1M Urea, pH 7.4. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.