Learn More
Enzo Life Sciences Ubiquitin activating kit (20 tests)
SDP
Supplier:  Enzo Life Sciences BMLUW0400A0001
Encompass_Preferred
For activation of ubiquitin for use in ubiquitinylation experiments. The kit provides the means of generating thioester linked, activated ubiquitin-E1 conjugates, utilizing the first step in the ubiquitin cascade, for investigation of ubiquitin activation, subsequent ubiquitin transfer to/interaction with E2 conjugating enzymes, and their use in the ubiquitinylation of E3 ligases and target substrate proteins. Ubiquitin antibody provided allows high sensitivity detection of ubiquitin and ubiquitin-conjugates.
Catalog No. 50-201-2249