Learn More
Enzo Life Sciences Ubiquitin, (agarose immobilized) (0.5ML)
SDP
Catalog No. 502012288
Encompass_Preferred
Formulation: Liquid. Suspension. Source: Ubiquitin is coupled to agarose, via the ε amino groups of its internal lysine residues, leaving the C-terminus free. The loading is approximately10mg protein/ml of settled resin. Long Term Storage: +4°C.
Catalog No. 50-201-2288 Supplier Enzo Life Sciences Supplier No. BMLUW86300500
Formulation: Liquid. Suspension. Source: Ubiquitin is coupled to agarose, via the ε amino groups of its internal lysine residues, leaving the C-terminus free. The loading is approximately10mg protein/ml of settled resin. Long Term Storage: +4°C.