Learn More
AdipoGen Ubiquitin aldehyde (human) (rec.) (50 µg)
SDP
Supplier:  AdipoGen SBBPS0031C050
Encompass_Preferred
Ubiquitin aldehyde (human) (rec.) (50 µg), AdipoGen Life Sciences. Protein. Human ubiquitin (aa1-76)with a C-terminal aldehyde. Source: E. coli. Formulation: Liquid. In 50mM NaOAc pH 5.0. Purity: >97% (LCMS). Ubiquitin is a small (8.5kDa) regulatory protein that has been found in almost all tissues of eukaryotic organisms. The addition of ubiquitin to a substrate protein is called ubiquitination or ubiquitylation. This product consists of a full-length human, mature ubiquitin polypeptide (amino acids 1-76), expressed in E.coli with a C-terminal warhead (Aldehyde). Ubiquitin aldehyde is a potent, irreversible and specific inhibitor of deubiquitinating enzymes (DUBs) based on a C-terminal electrophilic aldehyde group. Ubiquitin aldehyde can be used for activity profiling experiments and determining DUB inhibitor specificity.
Catalog No. 50-148-9537