Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHEMPEP INC Ubiquitin-AMC, 100 μg, Fluorogenic substrate for DUB activity assays, C388H636N106O119S, Molecular Weight 8722.2 g/mol, Fluorescence Excitation 380 nm, Emission 460 nm

Catalog No. NC2311940
Encompass

Ubiquitin-AMC is a highly sensitive fluorogenic substrate essential for measuring deubiquitinating enzyme (DUB) activity in biochemical assays. This versatile reagent, composed of ubiquitin conjugated to the AMC fluorophore, facilitates real-time monitoring of DUB activity by releasing a fluorescent signal. It is particularly advantageous for high-throughput screening and detailed kinetic studies in various cellular processes.

  • 100 μg quantity
  • Molecular formula: C388H636N106O119S
  • Molecular weight: 8722.2 g/mol
  • Fluorogenic substrate type
  • Target enzyme class: Deubiquitinating enzymes (DUBs)
  • Real-time monitoring capability
  • High sensitivity for biochemical research
  • Uses aqueous buffers for solubility
  • Storage temperature: -20°C or below
  • Avoid repeated freeze-thaw cycles
  • Purity of ≥95%
  • Synthetic source of ubiquitin
  • Supplied as lyophilized powder or solution in DMSO

Catalog No. NC2311940 Supplier CHEMPEP INC Supplier No. 302109
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ubiquitin-AMC is a highly sensitive fluorogenic substrate essential for measuring deubiquitinating enzyme (DUB) activity in biochemical assays. This versatile reagent, composed of ubiquitin conjugated to the AMC fluorophore, facilitates real-time monitoring of DUB activity by releasing a fluorescent signal. It is particularly advantageous for high-throughput screening and detailed kinetic studies in various cellular processes.

  • 100 μg quantity
  • Molecular formula: C388H636N106O119S
  • Molecular weight: 8722.2 g/mol
  • Fluorogenic substrate type
  • Target enzyme class: Deubiquitinating enzymes (DUBs)
  • Real-time monitoring capability
  • High sensitivity for biochemical research
  • Uses aqueous buffers for solubility
  • Storage temperature: -20°C or below
  • Avoid repeated freeze-thaw cycles
  • Purity of ≥95%
  • Synthetic source of ubiquitin
  • Supplied as lyophilized powder or solution in DMSO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.