Learn More
Enzo Life Sciences Ubiquitin-AMC (25 µg)
SDP
Catalog No. 502011956
Encompass_Preferred
Ubiquitin-AMC is a fluorogenic substrate for a wide range of deubiquitinylating enzymes (DUBs), including ubiquitin C-terminal hydrolases (UCHs) and ubiquitin specific proteases (USPs). It is a particularly useful reagent for the study of deubiquitinylating activity where detection sensitivity or continuous monitoring of activity is essential. Ubiquitin-AMC has been shown to be a sensitive substrate for UCH-L3 (Km = 0.039μM) (BML-UW9745) and for Isopeptidase-T (Km = 0.17-1.4μM) (BML-UW9690), and is particularly useful for studying deubiquitinylating activity where detection sensitivity or continuous monitoring of activity is essential. Purity: ≥95% (HPLC). Formulation: In DMSO. Source: Produced in E.coli.
Catalog No. 50-201-1956 Supplier Enzo Life Sciences Supplier No. BMLSE2110025
Ubiquitin-AMC is a fluorogenic substrate for a wide range of deubiquitinylating enzymes (DUBs), including ubiquitin C-terminal hydrolases (UCHs) and ubiquitin specific proteases (USPs). It is a particularly useful reagent for the study of deubiquitinylating activity where detection sensitivity or continuous monitoring of activity is essential. Ubiquitin-AMC has been shown to be a sensitive substrate for UCH-L3 (Km = 0.039μM) (BML-UW9745) and for Isopeptidase-T (Km = 0.17-1.4μM) (BML-UW9690), and is particularly useful for studying deubiquitinylating activity where detection sensitivity or continuous monitoring of activity is essential. Purity: ≥95% (HPLC). Formulation: In DMSO. Source: Produced in E.coli.