Learn More
Santa Cruz Biotechnology Ubiquitin Antibody (A-5) 200 µg/ml: sc-166553
Catalog No. NC3355045
Encompass
Ubiquitin Antibody (A-5) is a mouse monoclonal IgG2b κ, cited in 95 publications, provided at 200 µg/mlraised against amino acids 1-76 representing full length Ubiquitin of human originAnti-Ubiquitin Antibody (A-5) is recommended for detection of Ubiquitin and polyubiquitin of mouse, rat and human origin by WB, IP, IF, IHC(P) and ELISAAnti-Ubiquitin Antibody (A-5) is available conjugated to agarose for IP; HRP for WB, IHC(P) and ELISA; and to either phycoerythrin or FITC for IF, IHC(P) and FCM
Catalog No. NC3355045 Supplier Santa Cruz Biotechnology Supplier No. SC166553
Ubiquitin Antibody (A-5) is a mouse monoclonal IgG2b κ, cited in 95 publications, provided at 200 µg/mlraised against amino acids 1-76 representing full length Ubiquitin of human originAnti-Ubiquitin Antibody (A-5) is recommended for detection of Ubiquitin and polyubiquitin of mouse, rat and human origin by WB, IP, IF, IHC(P) and ELISAAnti-Ubiquitin Antibody (A-5) is available conjugated to agarose for IP; HRP for WB, IHC(P) and ELISA; and to either phycoerythrin or FITC for IF, IHC(P) and FCM