Learn More
Enzo Life Sciences Ubiquitin polyclonal antibody (50 µg)
SDP
Supplier:  Enzo Life Sciences ADISPA200D
Encompass_Preferred
Host: Rabbit. Immunogen: Native bovine Ubiquitin. UniProt: P0CG53. Source: Purified from rabbit serum. Species Reactivity: Human, Mouse, Rat, Yeast, Xenopus, Sheep, Rabbit, Porcine, Monkey, Hamster, Guinea pig, Fish, Drosophila, Dog, Chicken, Bovine. Applications: ICC, IHC (PS), WB. Recommended dilutions: Western Blot (1:5,000, ECL). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-667-424