Learn More
Santa Cruz Biotechnology UBIQUITIN ANTIBODY (P4D1)
Supplier:  Santa Cruz Biotechnology SC8017
Encompass
Ub (P4D1), mouse monoclonal IgG1; 200microgram/ml, epitope corresponding to amino acids 1-76 representing full length Ub of bovine origin, reactive with ubiquitin, poly-ubiquitin and ubiquitinated proteins broadly reactive with multiple spe
Catalog No. NC9831782