Learn More
Bioss Ubiquitin C-terminal Hydrolase
SDP
Catalog No. 5000065174
Encompass_Preferred
Ubiquitin C-terminal Hydrolase 1 (UCHL1) Primary Antibody Unconjugated Host Species Mouse Target Species Human Mouse Rat Bovine Pig Clonality Monoclonal Applications WB IHC BH7Concentration Lot Dependent Size 100 ul Storage Buffer PBS 50% glycerol and 5 mm NaN3
Catalog No. 50-000-65174 Supplier Bioss Supplier No. BSM70306M
Ubiquitin C-terminal Hydrolase 1 (UCHL1) Primary Antibody Unconjugated Host Species Mouse Target Species Human Mouse Rat Bovine Pig Clonality Monoclonal Applications WB IHC BH7Concentration Lot Dependent Size 100 ul Storage Buffer PBS 50% glycerol and 5 mm NaN3