Learn More
Enzo Life Sciences Ubiquitin (human), (recombinant) (His-tag) (1 mg)
SDP
Supplier:  Enzo Life Sciences BMLUW86100001
Encompass_Preferred
His-Ubiquitin is activated by E1 and is a competent substrate for the E2 and E3 enzymes. His-Ubiquitin is activated by E1 and is a competent substrate for the E2 and E3 enzymes. Formulation: Lyophilized. Source: Produced in E. coli. Long Term Storage: -20°C.
Catalog No. 50-201-2286