Learn More
AdipoGen Ubiquitin (human) (rec.) (Cy5) (50 µg)
SDP
Supplier:  AdipoGen SBBTR0015C050
Encompass_Preferred
Ubiquitin (human) (rec.) (Cy5) (50 µg), AdipoGen Life Sciences. Protein. Human ubiquitin (aa1-76)is site-specifically conjugated to a single Cyanine 5 (Cy5) moiety. All lysines are available along with a fully functional C-terminus, making it ideal for chain formation using the E1-E2 and E3 conjugation cascade. Source: E. coli. Formulation: Liquid. In 50mM HEPES pH 7.5, 50mM sodium chloride. Purity: >98% (LCMS). Ubiquitination, the conjugation of ubiquitin to other proteins through a covalent bond between its C-terminal glycine and the epsilon-amino group of lysine residues or the alpha-amino group of an N- terminal methionine onto proteins is essential for many cellular processes primarily linked to protein degradation. This Europium Cryptate Ubiquitin is ideal for measuring Ub chain conjugation or deconjugation using TR-FRET measurement. Its TR-FRET pair is Europium-Cryptate-Ubiquitin.
Catalog No. 50-148-9542