Learn More
Cayman Chemical Ubiquitin Isopeptidase Inhibitor I, Purity: ≥98%, CAS: 108477-18-5, 25 mg
SDP
Catalog No. 502983557
Encompass_Preferred
Induces caspase activation and apoptosis (IC50 = ~1.7 µM) through a Bcl-2-dependent and apoptosome-independent mitochondrial pathway that selectively inhibits ubiquitin isopeptidase activity (IC50 = ~ 30 µM)
Catalog No. 50-298-3557 Supplier Cayman Chemical Supplier No. 2100625 MG
Induces caspase activation and apoptosis (IC50 = ~1.7 µM) through a Bcl-2-dependent and apoptosome-independent mitochondrial pathway that selectively inhibits ubiquitin isopeptidase activity (IC50 = ~ 30 µM)