Learn More
Cayman Chemical UbIquItIn IsoPEPIDSE InHBI 5mg
SDP
Catalog No. 502984153
Encompass_Preferred
Induces caspase activation and apoptosis (IC50 = ~1.7 µM) through a Bcl-2-dependent and apoptosome-independent mitochondrial pathway that selectively inhibits ubiquitin isopeptidase activity (IC50 = ~ 30 µM)
Catalog No. 50-298-4153 Supplier Cayman Chemical Supplier No. 210065 MG
Induces caspase activation and apoptosis (IC50 = ~1.7 µM) through a Bcl-2-dependent and apoptosome-independent mitochondrial pathway that selectively inhibits ubiquitin isopeptidase activity (IC50 = ~ 30 µM)