Learn More
Enzo Life Sciences Ubiquitin monoclonal antibody (P4D1) (HRP conjugate) (25 µl)
SDP
Supplier:  Enzo Life Sciences BMLPW09350025
Encompass_Preferred
Clone: P4D1. Isotype: IgG1. Immunogen: Denatured bovine ubiquitin. Source: Purified from tissue culture supernatant. HRP conjugated chemically in vitro. Species reactivity: Species independent. Specificity: Recognizes mono- and poly-ubiquitin protein conjugates, free polyubiquitin chains and free ubiquitin. Applications: WB. Purity: Ion exchange purified. Formulation: Liquid. In PBS containing 0.01% sodium azide.
Catalog No. 50-201-1556