Learn More
Enzo Life Sciences Ubiquitin monoclonal antibody (P4G7-H11) (200µg)
SDP
Supplier:  Enzo Life Sciences ADISPA203F
Encompass_Preferred
Clone: P4G7-H11. Host: Mouse. Isotype: IgG1. Immunogen: Native bovine Ubiquitin. UniProt ID: P0CG53. Source: Purified from ascites. Species reactivity: Human, Mouse, Rat, Rabbit, Hamster, Bovine. Applications: Flow Cytometry, IHC (PS), WB. Recommended Dilutions: Flow Cytometry (10µg/ml) Western Blot (1:1,000, colorimetric) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7415