Learn More
R&D Systems™ Ubiquitin-Propargylamide Protein
Description
Specifications
Specifications
| Formulation | Lyophilized from a solution in Acetonitrile, Acetic Acid, deionized water. |
| Molecular Weight (g/mol) | 8.5 kDa |
| Name | Ubiquitin Propargylamide |
| Quantity | 50μg |
| Source | Chemically Synthesized Ubiquitin protein |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | PA-Tag |
| Recombinant | Recombinant |
| Purity or Quality Grade | 95%, by HPLC. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.